missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-3-fluoropyridine, TRC

Product Code. 30384752
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384752 500 mg
30372049 100 mg
30369765 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384752 Supplier Toronto Research Chemicals Supplier No. TRCD425738500MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41404-59-5
Molecular Formula C5H2Br2FN
Molecular Weight (g/mol) 254.88
SMILES Fc1ccc(Br)nc1Br

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H2Br2FN/c6-4-2-1-3(8)5(7)9-4/h1-2H
Formula Weight 252.8538
Chemical Name or Material 2,6-Dibromo-3-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.