Learn More
2,6-dibromo-3-chloro-4-fluoroaniline, Maybridge
Brand: Maybridge BTB04528EB
Code : 31
Additional Details : CAS Number : 175135-09-8 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
175135-09-8 | |
303.353 | |
3-chloro-2,6-dibromo-4-fluoroaniline, 2,6-dibromo-3-chloro-4-fluorobenzenamine, 1-chloro-2,6-dibromo-4-fluoroaniline, benzenamine, 2,6-dibromo-3-chloro-4-fluoro, benzenamine,2,6-dibromo-3-chloro-4-fluoro, pubchem2948, acmc-1c6hf, 2,6-dibromo-3-chloro-4-fluorophenylamine | |
2,6-dibromo-3-chloro-4-fluoroaniline |
C6H3Br2ClFN | |
PRZBEZGEEPNKRO-UHFFFAOYSA-N | |
2736480 | |
C1=C(C(=C(C(=C1Br)N)Br)Cl)F |
Specifications
25g | |
Amber Glass Bottle | |
C6H3Br2ClFN | |
PRZBEZGEEPNKRO-UHFFFAOYSA-N | |
2,6-dibromo-3-chloro-4-fluoroaniline | |
2736480 | |
97% |
2, 6-dibromo-3-chloro-4-fluoroaniline | |
175135-09-8 | |
3-chloro-2,6-dibromo-4-fluoroaniline, 2,6-dibromo-3-chloro-4-fluorobenzenamine, 1-chloro-2,6-dibromo-4-fluoroaniline, benzenamine, 2,6-dibromo-3-chloro-4-fluoro, benzenamine,2,6-dibromo-3-chloro-4-fluoro, pubchem2948, acmc-1c6hf, 2,6-dibromo-3-chloro-4-fluorophenylamine | |
C1=C(C(=C(C(=C1Br)N)Br)Cl)F | |
303.353 | |
303.36 |