missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminotoluene, 98%

Product Code. 10418900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10418900 25 g Glass bottle 25g
10519443 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10418900 Supplier Thermo Scientific Acros Supplier No. 164680250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 823-40-5 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-40-5
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007800
InChI Key RLYCRLGLCUXUPO-UHFFFAOYSA-N
Synonym 2,6-diaminotoluene, 2,6-toluenediamine, 1,3-benzenediamine, 2-methyl, toluene-2,6-diamine, 2,6-tolylenediamine, 2,6-toluylenediamine, 2,6-diamino-1-methylbenzene, 2-methyl-1,3-benzenediamine, 2-methyl-1,3-phenylenediamine, 2-methyl-m-phenylenediamine
PubChem CID 13205
ChEBI CHEBI:76288
IUPAC Name 2-methylbenzene-1,3-diamine
SMILES CC1=C(N)C=CC=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 102.0°C to 107.0°C
Color Black or Brown to Gray
Boiling Point 289.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NH2)2
Quantity 25 g
Beilstein 13,148
Solubility Information Solubility in water: 60g/L water (15°C).
Formula Weight 122.17
Percent Purity 98%
Physical Form Pellets, Powder or Chunks
Chemical Name or Material 2, 6-Diaminotoluene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,6-Diaminotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.