missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopyridine, 98%

Product Code. 10263563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10263563 25 g Glass bottle 25g
10469020 100 g Glass bottle 100g
10530201 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10263563 Supplier Thermo Scientific Acros Supplier No. 112380250
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141-86-6 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-86-6
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006329
InChI Key VHNQIURBCCNWDN-UHFFFAOYSA-N
Synonym 2,6-diaminopyridine, 2,6-pyridinediamine, 2,6-diamino pyridine, pyridine, 2,6-diamino, dap amine, pyridine-2,6-diyldiamine, unii-k89ab8ckg6, ccris 6682, k89ab8ckg6, dsstox_cid_20127
PubChem CID 8861
IUPAC Name pyridine-2,6-diamine
SMILES C1=CC(=NC(=C1)N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 115°C to 123°C
Color Beige to Brown-Gray
Boiling Point 285°C
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)(NH2)2
Quantity 25 g
Beilstein 22, I, 647
Solubility Information Solubility in water: 9.9g/100 mL (20°C)
Formula Weight 109.13
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2, 6-Diaminopyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Diaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2,6-Pyridinediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.