missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopyridine, 98%

Product Code. 11445033
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445033 500 g 500g
11435033 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445033 Supplier Thermo Scientific Alfa Aesar Supplier No. A12295.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141-86-6 | C5H7N3 | 109.132 g/mol

2,6-Diaminopyridine, used as a pharmaceutical intermediate and a hair dye coupler in oxidation/permanent formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diaminopyridine, used as a pharmaceutical intermediate and a hair dye coupler in oxidation/permanent formulations.

Solubility
Soluble in water 180 g/L @ 20°C, acetone, ethanol, methanol, isopropanol, ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-86-6
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD00006329
InChI Key VHNQIURBCCNWDN-UHFFFAOYSA-N
Synonym 2,6-diaminopyridine, 2,6-pyridinediamine, 2,6-diamino pyridine, pyridine, 2,6-diamino, dap amine, pyridine-2,6-diyldiamine, unii-k89ab8ckg6, ccris 6682, k89ab8ckg6, dsstox_cid_20127
PubChem CID 8861
IUPAC Name pyridine-2,6-diamine
SMILES C1=CC(=NC(=C1)N)N

Specifications

Melting Point 118°C to 121°C
Boiling Point 288°C
Flash Point 155°C (311°F)
Odor Unpleasant
Quantity 500 g
UN Number UN2671
Beilstein 108513
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2,6-Diaminopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.