missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diaminopimelic Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2,6-Diaminopimelic acid |
| CAS | 583-93-7 |
| Molecular Formula | C7 H14 N2 O4 |
| Quantity | 1 g |
| Synonym | α,α'-Diaminopimelic acid |
| InChI Formula | InChI=1S/C7H14N2O4/c8-4(6(10)11)2-1-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13) |
| SMILES | NC(CCCC(N)C(=O)O)C(=O)O |
| IUPAC Name | 2,6-diaminoheptanedioic acid |
| Molecular Weight (g/mol) | 190.2 |
| Formula Weight | 190.0954 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction