missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diaminopimelic Acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2,6-Diaminopimelic acid |
| CAS | 583-93-7 |
| Molecular Formula | C7 H14 N2 O4 |
| Quantity | 100 mg |
| Synonym | α,α'-Diaminopimelic acid |
| InChI Formula | InChI=1S/C7H14N2O4/c8-4(6(10)11)2-1-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13) |
| SMILES | NC(CCCC(N)C(=O)O)C(=O)O |
| IUPAC Name | 2,6-diaminoheptanedioic acid |
| Molecular Weight (g/mol) | 190.2 |
| Formula Weight | 190.0954 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering