missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diacetylpyridine, 99%

Product Code. 10666501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10666501 5 g Glass bottle 5g
10656501 1 g Glass bottle 1g
10125683 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10666501 Supplier Thermo Scientific Acros Supplier No. 330860050
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1129-30-2 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-30-2
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00006304
InChI Key BEZVGIHGZPLGBL-UHFFFAOYSA-N
Synonym 2,6-diacetylpyridine, 1,1'-pyridine-2,6-diyl diethanone, 1-6-acetylpyridin-2-yl ethanone, pyridine-2,6-diacetyl, 2,6-diacetyl pyridine, 1-6-acetyl-pyridin-2-yl-ethanone, ethanone, 1,1'-2,6-pyridinediyl bis, 1-6-acetylpyridin-2-yl ethan-1-one, 2,6diacetylpyridine, pubchem3144
PubChem CID 70790
IUPAC Name 1-(6-acetylpyridin-2-yl)ethanone
SMILES CC(=O)C1=CC=CC(=N1)C(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 78°C to 82°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)(COCH3)2
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform,dimethylsulfoxide
Formula Weight 163.18
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 2, 6-Diacetylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Diacetylpyridine
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.