missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butylphenol, TRC

Product Code. 30373637
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373637 Supplier Toronto Research Chemicals Supplier No. TRCD4945931G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Di-tert-butylphenol
CAS 128-39-2
Molecular Formula C14 H22 O
Quantity 1 g
Synonym Phenol, 2,6-di-tert-butyl- (8CI), 2,6-Bis(1,1-dimethylethyl)phenol, 2,6-Bis(tert-butyl)phenol, 2,6-Di-(1,1-Dimethylethyl)phenol, 2,6-Di-t-butylphenol, 2,6-Di-tert-butyl-4-phenol, 2,6-Di-tert-butylhydroxybenzene, 2,6-Di-tert-butylphenol, 2,6-tert-Butylphenol, AN 701, Agidol 0, Aladdin 1138077, Antioxidant T 502, Ethanox 4701, Ethanox 4733, Ethyl 701, Ethyl AN 701, Hitec 4701, Isonox 103, KY 02, Kerobit TP 26, NSC 49175, Na Lube AO 242, T 502, T 502 (antioxidant)
InChI Formula InChI=1S/C14H22O/c1-13(2,3)10-8-7-9-11(12(10)15)14(4,5)6/h7-9,15H,1-6H3
SMILES CC(C)(C)c1cccc(c1O)C(C)(C)C
IUPAC Name 2,6-ditert-butylphenol
Molecular Weight (g/mol) 206.32
Formula Weight 206.1671
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.