missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butylphenol, 99%

Product Code. p-3736151 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10513104

Brand: Thermo Scientific Acros 113002500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128-39-2 | C14H22O | 206.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-39-2
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.33
MDL Number MFCD00008820
InChI Key DKCPKDPYUFEZCP-UHFFFAOYSA-N
Synonym 2,6-di-tert-butylphenol, 2,6-di-t-butylphenol, 2,6-bis tert-butyl phenol, 2,6-bis 1,1-dimethylethyl phenol, phenol, 2,6-bis 1,1-dimethylethyl, 2,6 di-tert-butylphenol, ethanox 701, isonox 103, ethyl 701, ethyl an 701
PubChem CID 31405
IUPAC Name 2,6-ditert-butylphenol
SMILES CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 35°C to 38°C
Color White to Yellow
Boiling Point 253°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula [(CH3)3C]2C6H3OH
Quantity 250 g
Beilstein 06, III, 2061
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and benzene
Formula Weight 206.33
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2, 6-Di-tert-butylphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Di-tert-butylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2,6-Di-tert-butylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.