missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butylphenol, 99%, Thermo Scientific Chemicals

Catalog Number 10654952
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
128-39-2
C14H22O
206.33
MFCD00008820
DKCPKDPYUFEZCP-UHFFFAOYSA-N
2,6-di-tert-butylphenol, 2,6-di-t-butylphenol, 2,6-bis tert-butyl phenol, 2,6-bis 1,1-dimethylethyl phenol, phenol, 2,6-bis 1,1-dimethylethyl, 2,6 di-tert-butylphenol, ethanox 701, isonox 103, ethyl 701, ethyl an 701
31405
2,6-ditert-butylphenol
CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C
This item is not returnable. View return policy
128-39-2
C14H22O
206.33
MFCD00008820
DKCPKDPYUFEZCP-UHFFFAOYSA-N
2,6-di-tert-butylphenol, 2,6-di-t-butylphenol, 2,6-bis tert-butyl phenol, 2,6-bis 1,1-dimethylethyl phenol, phenol, 2,6-bis 1,1-dimethylethyl, 2,6 di-tert-butylphenol, ethanox 701, isonox 103, ethyl 701, ethyl an 701
31405
2,6-ditert-butylphenol
CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 35°C to 38°C
Color White to Yellow
Boiling Point 253°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula [(CH3)3C]2C6H3OH
Quantity 1 kg
Beilstein 06, III, 2061
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and benzene
Formula Weight 206.33
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2, 6-Di-tert-butylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'