missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di(tert-butyl)pyridine, 97%, Thermo Scientific™, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Density | 0.8520g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | (C5H3N[C(CH3)3]2 |
Quantity | 1 g |
Formula Weight | 191.32 |
Percent Purity | 97% |
Chemical Name or Material | 2, 6-Di(tert-butyl)pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'