missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylpyridine, 98%

Product Code. 10558333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10558333 25 g Glass bottle 25g
10175892 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10558333 Supplier Thermo Scientific Acros Supplier No. 224520250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 38222-83-2 | C14H23N | 205.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38222-83-2
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00006305
InChI Key HVHZEKKZMFRULH-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylpyridine, dtbmp, pyridine, 2,6-bis 1,1-dimethylethyl-4-methyl, 2,6-di-t-butyl-4-methylpyridine, 2,6-ditert-butyl-4-methyl-pyridine, 2,6-bis tert-butyl-4-methylpyridine, pubchem10244, acmc-209iyr, ksc490s2n, 2,6-di-t-butyl 4-methylpyridine
PubChem CID 98898
IUPAC Name 2,6-ditert-butyl-4-methylpyridine
SMILES CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color Colorless to Yellow
Boiling Point 233°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3C]2C5H2NCH3
Refractive Index 1.4760 to 1.478
Quantity 25 g
Beilstein 20, V,6, 190
Fieser 09,141; 10,123; 11,160; 16,110
Merck Index 15, 3042
Formula Weight 205.34
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2, 6-Di-tert-butyl-4-methylpyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Di-tert-butyl-4-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.