missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)pyridine, 97%

Product Code. 11398746
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398746 250 mg 250mg
11388746 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398746 Supplier Thermo Scientific Alfa Aesar Supplier No. L19446.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 455-00-5 | C7H3F6N | 215.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-00-5
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD00236675
InChI Key YPDVFTXBESQIPJ-UHFFFAOYSA-N
PubChem CID 589864
IUPAC Name 2,6-bis(trifluoromethyl)pyridine
SMILES C1=CC(=NC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 55°C to 58°C
Boiling Point 82°C (18 mmHg)
Flash Point 67°C (153°F)
Quantity 250 mg
Beilstein 188806
Synonym 2,6-bis trifluoromethyl pyridine, 2,6-bis trifluoromethyl-pyridine, pubchem7696, acmc-209k2t, pyridine,2,6-bis trifluoromethyl, pyridine, 2,6-bis trifluoromethyl
Formula Weight 215.1
Percent Purity 97%
Chemical Name or Material 2,6-Bis(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.