missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%

Product Code. 11384316
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384316 1 g 1g
11304326 250 mg 250mg
11394316 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384316 Supplier Thermo Scientific Alfa Aesar Supplier No. L19952.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 681812-07-7 | C8H5BF6O2 | 257.926 g/mol

2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681812-07-7
Molecular Formula C8H5BF6O2
Molecular Weight (g/mol) 257.926
MDL Number MFCD04039321
InChI Key WAMPGNNEOZGBHR-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid
PubChem CID 2783239
IUPAC Name [2,6-bis(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O

Specifications

Melting Point ∼150°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 257.93
Percent Purity 97%
Chemical Name or Material 2,6-Bis(trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.