missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7058525
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
681812-07-7
C8H5BF6O2
257.926
MFCD04039321
WAMPGNNEOZGBHR-UHFFFAOYSA-N
2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid
2783239
[2,6-bis(trifluoromethyl)phenyl]boronic acid
B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O
This item is not returnable. View return policy
681812-07-7
C8H5BF6O2
257.926
MFCD04039321
WAMPGNNEOZGBHR-UHFFFAOYSA-N
2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid
2783239
[2,6-bis(trifluoromethyl)phenyl]boronic acid
B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O

2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼150°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 257.93
Percent Purity 97%
Chemical Name or Material 2,6-Bis(trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'