missing translation for 'onlineSavingsMsg'
Learn More

2-[6-(Acetylphenylamino)-1,3,5-hexatrienyl]-1,1-dimethyl-3-sulfobutyl-1H-benz[e]indolium Inner Salt, TRC

Product Code. 30005161
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005161 1 g 1g
30010533 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005161 Supplier Toronto Research Chemicals Supplier No. TRCA1879401G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63450-66-8
Molecular Formula C32 H34 N2 O4 S
Molecular Weight (g/mol) 542.69
IUPAC Name 4-[2-[(1E,3E,5E)-6-(N-acetylanilino)hexa-1,3,5-trienyl]-1,1-dimethyl-benzo[e]indol-3-ium-3-yl]butane-1-sulfonate
SMILES CC(=O)N(\C=C\C=C\C=C\C1=[N+](CCCCS(=O)(=O)[O-])c2ccc3ccccc3c2C1(C)C)c4ccccc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C32H34N2O4S/c1-25(35)33(27-16-7-6-8-17-27)22-12-5-4-9-19-30-32(2,3)31-28-18-11-10-15-26(28)20-21-29(31)34(30)23-13-14-24-39(36,37)38/h4-12,15-22H,13-14,23-24H2,1-3H3
Formula Weight 542.2239
Chemical Name or Material 2-[6-(Acetylphenylamino)-1,3,5-hexatrienyl]-1,1-dimethyl-3-sulfobutyl-1H-benz[e]indolium Inner Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.