missing translation for 'onlineSavingsMsg'
Learn More

2,6,10,14-Tetramethylpentadecane, 95%

Product Code. 10240753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10240753 250 mL Glass bottle 250mL
10427150 5 mL Glass bottle 5mL
10052872 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10240753 Supplier Thermo Scientific Acros Supplier No. 138462500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 1921-70-6 | C19H40 | 268.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1921-70-6
Molecular Formula C19H40
Molecular Weight (g/mol) 268.51
InChI Key XOJVVFBFDXDTEG-UHFFFAOYSA-N
Synonym pristane, norphytane, bute hydrocarbon, norphytan, pristan, pentadecane, 2,6,10,14-tetramethyl, meso-pristane, 2,6,10,10-tetramethylpentadecane, meso-form, norphytane, robuoy
PubChem CID 15979
ChEBI CHEBI:53181
IUPAC Name 2,6,10,14-tetramethylpentadecane
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.7800g/mL
Boiling Point 296°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)(CH2)3CH(CH3)2
Refractive Index 1.4369 to 1.4389
Quantity 250 mL
Merck Index 15,787
Specific Gravity 0.78
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,petroleum ether,benzene,,chloroform and carbon tetrachloride
Viscosity 5 mPa.s (25°C)
Formula Weight 268.51
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2,6,10,14-Tetramethylpentadecane, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6,10,14-Tetramethylpentadecane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2,6,10,14-Tetramethylpentadecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.