missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vindesine N-(3-Hydroxypropyl)amide-13C3,15N, TRC
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | Vindesine N-(3-Hydroxypropyl)amide-13C3,15N |
| Molecular Formula | 13C3 C43 H61 15N N4 O8 |
| Quantity | 2.5 mg |
| Synonym | Vindesine HPA-13C3,15N, (3S,5S,7S,9S)-Methyl 5-Ethyl-9-((3aR,3a1R,4R,5S,5aR,10bR)-3a-ethyl-4,5-dihydroxy-5-((3-hydroxypropyl)carbamoyl)-8-methoxy-6-methyl-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazol-9-yl)-5-hydroxy-2,4,5,6,7,8,9,10-octahydro-1H-3,7-methano[1]azacycloundecino[5,4-b]indole-9-carboxylate-13C3,15N |
| InChI Formula | InChI=1S/C46H61N5O8/c1-6-42(56)24-28-25-45(41(55)59-5,36-30(14-19-50(26-28)27-42)29-12-8-9-13-33(29)48-36)32-22-31-34(23-35(32)58-4)49(3)38-44(31)16-20-51-18-10-15-43(7-2,37(44)51)39(53)46(38,57)40(54)47-17-11-21-52/h8-10,12-13,15,22-23,28,37-39,48,52-53,56-57H,6-7,11,14,16-21,24-27H2,1-5H3,(H,47,54)/t28-,37+,38-,39-,42+,43-,44-,45+,46+/m1/s1/i11+1,17+1,21+1,47+1 |
| SMILES | CC[C@]1(O)C[C@H]2C[N@](CCc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C)[C@H]7[C@](O)([C@H](O)[C@]8(CC)C=CCN9CC[C@]67[C@H]89)C(=O)[15NH][13CH2][13CH2][13CH2]O)C1 |
| IUPAC Name | methyl (1S,13S,15S,17S)-17-ethyl-13-[(1R,9R,10S,11R,12R,19R)-12-ethyl-10,11-dihydroxy-10-(3-hydroxy(1,2,3-13C3)propylcarbamoyl)-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.01,9.02,7.016,19]nonadeca-2,4,6,13-tetraen-4-yl]-17-hydroxy-1,11-diazatetracyclo[13.3.1.04,12.05,10]nonadeca-4(12),5,7,9-tetraene-13-carboxylate |
| Molecular Weight (g/mol) | 815.977 |
| Formula Weight | 815.459 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction