missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vardenafil Oxopiperazine-D6 (Impurity), TRC
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
Specifications
Specifications
| Chemical Name or Material | Vardenafil Oxopiperazine-D6 (Impurity) |
| Molecular Formula | C21 D6 H20 N6 O5 S |
| Quantity | 2.5 mg |
| Synonym | 2-[2-Ethoxy-5-[(3-oxo-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-imidazo[5,1-f][1,2,4]triazin-4(1H)-one-D6, 4-[[3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]-piperazinone-D6 |
| InChI Formula | InChI=1S/C21H26N6O5S/c1-4-6-17-23-13(3)19-21(29)24-20(25-27(17)19)15-11-14(7-8-16(15)32-5-2)33(30,31)26-10-9-22-18(28)12-26/h7-8,11H,4-6,9-10,12H2,1-3H3,(H,22,28)(H,24,25,29)/i9D2,10D2,12D2 |
| SMILES | [2H]C1([2H])NC(=O)C([2H])([2H])N(C1([2H])[2H])S(=O)(=O)c2ccc(OCC)c(c2)C3=NC(=O)c4c(C)nc(CCC)n4N3 |
| IUPAC Name | 2-[2-ethoxy-5-(2,2,3,3,6,6-hexadeuterio-5-oxopiperazin-1-yl)sulfonylphenyl]-5-methyl-7-propyl-1H-imidazo[5,1-f][1,2,4]triazin-4-one |
| Molecular Weight (g/mol) | 480.57 |
| Formula Weight | 480.206 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction