missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophol-d4, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Tryptophol-d4 |
| CAS | 75238-45-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 D4 H7 N O |
| Quantity | 2.5 mg |
| Synonym | 1H-Indole-3-ethan-alpha,alpha,beta,beta-d4-ol |
| InChI Formula | InChI=1S/C10H11NO/c12-6-5-8-7-11-10-4-2-1-3-9(8)10/h1-4,7,11-12H,5-6H2/i5D2,6D2 |
| SMILES | [2H]C([2H])(O)C([2H])([2H])c1c[nH]c2ccccc12 |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(1H-indol-3-yl)ethanol |
| Molecular Weight (g/mol) | 165.225 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction