missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiamethoxam-d4, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | ThiamethoxamD4(oxadiazineD4) |
| CAS | 1331642-98-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 2H4 H6 Cl N5 O3 S |
| Quantity | 2.5 mg |
| Synonym | 4H-1,3,5-Oxadiazin-(2,2,6,6-D4)-4-imine','3-[(2-chloro-5-thiazolyl)methyl]tetrahydro-5-methyl-N-nitro- |
| InChI Formula | InChI=1S/C8H10ClN5O3S/c1-12-4-17-5-13(8(12)11-14(15)16)3-6-2-10-7(9)18-6/h2H,3-5H2,1H3/i4D2,5D2 |
| SMILES | [2H]C1([2H])OC([2H])([2H])N(Cc2cnc(Cl)s2)C(=N[N+](=O)[O-])N1C |
| IUPAC Name | N-[3-[(2-chloro-1,3-thiazol-5-yl)methyl]-2,2,6,6-tetradeuterio-5-methyl-1,3,5-oxadiazinan-4-ylidene]nitramide |
| Molecular Weight (g/mol) | 295.74 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction