missing translation for 'onlineSavingsMsg'
Learn More

(E)-Tamoxifen, TRC

Product Code. 30005804
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005804 2.5 mg 2.5mg
30011399 10 mg 10mg
30004636 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005804 Supplier Toronto Research Chemicals Supplier No. TRCT0060102.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13002-65-8
Molecular Formula C26 H29 N O
Molecular Weight (g/mol) 371.51
Synonym Ethanamine, 2-[4-[(1E)-1,2-diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethyl-, Ethanamine, 2-[4-(1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethyl-, (E)-, Ethanamine, 2-[4-[(1E)-1,2-diphenyl-1-butenyl]phenoxy]-N,N-dimethyl- (9CI), Ethylamine, 2-[p-(1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethyl-, (E)- (8CI), (E)-Tamoxifen, 2-[4-[(1E)-1,2-Diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethylethanamine, cis-Tamoxifen
IUPAC Name 2-[4-[(E)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
SMILES CC\C(=C(\c1ccccc1)/c2ccc(OCCN(C)C)cc2)\c3ccccc3

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C26H29NO/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3/h5-18H,4,19-20H2,1-3H3/b26-25+
Formula Weight 371.2249
Percent Purity >95%
Chemical Name or Material Tamoxifen E-isomer
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.