missing translation for 'onlineSavingsMsg'
Learn More

9,14-Deepoxy-15-deoxo-14-deoxy-9,15-epoxy-15-hydroxy-14-oxorapamycin (>80%), TRC

Product Code. 30010558
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010558 2.5 mg 2.5mg
30134489 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010558 Supplier Toronto Research Chemicals Supplier No. TRCD2285882.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150821-39-9
Molecular Formula C51H79NO13
Molecular Weight (g/mol) 914.17
Synonym Rapamycin, 9,14-deepoxy-15-deoxo-14-deoxy-9,15-epoxy-15-hydroxy-14-oxo-, 9,14-Deepoxy-15-deoxo-14-deoxy-9,15-epoxy-15-hydroxy-14-oxorapamycin, (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,28RS,34aS)-9,10,12,13,14,21,22,23,24,25,26,28,32,33,34,34a-Hexadecahydro-9,28-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,28-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,27,29(4H,6H,31H)-pentone
SMILES CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(/C)\[C@H](C[C@@H]3CC[C@@H](C)C(=O)C(O)(O3)C(=O)N4CCCC[C@H]4C(=O)O2)OC)CC[C@H]1O

Specifications

Analyte or Component Names 9,14-Deepoxy-15-deoxo-14-deoxy-9,15-epoxy-15-hydroxy-14-oxorapamycin
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Pharmaceutical
Quantity 2.5 mg
Formula Weight 913.5551
Chemical Name or Material 9,14-Deepoxy-15-deoxo-14-deoxy-9,15-epoxy-15-hydroxy-14-oxorapamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.