missing translation for 'onlineSavingsMsg'
Learn More

6-Hexadecanoylamino-4-methylumbelliferyl Beta-D-Galactopyranoside, TRC

Product Code. 30008495
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008495 2.5 mg 2.5mg
30132417 25 mg 25mg
30010761 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008495 Supplier Toronto Research Chemicals Supplier No. TRCH2909802.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94452-17-2
Molecular Formula C32H49NO9
Molecular Weight (g/mol) 591.73
Synonym N-[7-(beta-D-Galactopyranosyloxy)-4-methyl-2-oxo-2H-1-benzopyran-6-yl]- hexadecanamide, HMGal;
IUPAC Name N-[4-methyl-2-oxo-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-6-yl]hexadecanamide
SMILES CCCCCCCCCCCCCCCC(=O)Nc1cc2C(=CC(=O)Oc2cc1O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C32H49NO9/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-27(35)33-23-18-22-21(2)17-28(36)40-24(22)19-25(23)41-32-31(39)30(38)29(37)26(20-34)42-32/h17-19,26,29-32,34,37-39H,3-16,20H2,1-2H3,(H,33,35)/t26-,29+,30+,31-,32-/m1/s1
Formula Weight 591.3407
Percent Purity >95%
Chemical Name or Material 6-Hexadecanoylamino-4-methylumbelliferyl beta-D-Galactopyranoside
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.