missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Benzenetricarboxylic Acid 1,4-Bis(2-ethylhexyl) Ester, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | 1,2,4-Benzenetricarboxylic acid, 1,4-bis(2-ethylhexyl) ester |
| CAS | 63468-10-0 |
| Molecular Formula | C25H37O6- |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C25H38O6/c1-5-9-11-18(7-3)16-30-24(28)20-13-14-21(22(15-20)23(26)27)25(29)31-17-19(8-4)12-10-6-2/h13-15,18-19H,5-12,16-17H2,1-4H3,(H,26,27)/p-1 |
| SMILES | CCCCC(CC)COC(=O)C1=CC(=C(C=C1)C(=O)OCC(CC)CCCC)C(=O)[O-] |
| IUPAC Name | 2,5-bis(2-ethylhexoxycarbonyl)benzoate |
| Molecular Weight (g/mol) | 433.565 |
| Formula Weight | 433.259 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction