missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Chemical 2.5LT Ethanol 96% V/V 2.5L. Ar, 2.5LBP EP USP
Brand: Fisher Chemical E/0555/PC17
2.5LT Ethanol 96% V/V 2.5L. Ar, 2.5LBP EP USP
This item is not returnable.
View return policy
Description
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethanol (96% v/v)
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction