missing translation for 'onlineSavingsMsg'
Learn More

2(5H)-Furanone, 96%

Product Code. 11347926
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11347926

Brand: Thermo Scientific Alfa Aesar L13142.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 497-23-4 | C4H4O2 | 84.07 g/mol

2(5H)-Furanone is employed in the preparation of 5-substituted 2(5H) furanones (gamma-butenolides) catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts through direct aldol reaction with aromatic aldehydes. Further, it is used in the synthesis of lignans by Michael addition reactions. In addition to this, it is used in three-component Michael-Aldol reactions with an aldehyde and thiolate or carbanion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2(5H)-Furanone is employed in the preparation of 5-substituted 2(5H) furanones (gamma-butenolides) catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts through direct aldol reaction with aromatic aldehydes. Further, it is used in the synthesis of lignans by Michael addition reactions. In addition to this, it is used in three-component Michael-Aldol reactions with an aldehyde and thiolate or carbanion.

Solubility
Immiscible with water.

Notes
Light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-23-4
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00005376
InChI Key VIHAEDVKXSOUAT-UHFFFAOYSA-N
Synonym 2 5h-furanone, furan-2 5h-one, butenolide, 2-butenolide, gamma-crotonolactone, 2-buten-4-olide, isocrotonolactone, 2-oxo-2,5-dihydrofuran, crotonolactone, 2-5h-furanone
PubChem CID 10341
ChEBI CHEBI:38118
IUPAC Name 2H-furan-5-one
SMILES O=C1OCC=C1

Specifications

Melting Point 3°C to 4°C
Density 1.197
Boiling Point 212°C to 214°C
Flash Point 101°C (213°F)
Refractive Index 1.469
Quantity 25 g
Beilstein 383585
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 84.07
Percent Purity 98%
Chemical Name or Material 2(5H)-Furanone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.