missing translation for 'onlineSavingsMsg'
Learn More

2(5H)-Furanone, 95%

Product Code. 11936171 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11936171

Brand: Thermo Scientific Acros 376530010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 497-23-4 | C4H4O2 | 84.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-23-4
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00005376
InChI Key VIHAEDVKXSOUAT-UHFFFAOYSA-N
Synonym 2 5h-furanone, furan-2 5h-one, butenolide, 2-butenolide, gamma-crotonolactone, 2-buten-4-olide, isocrotonolactone, 2-oxo-2,5-dihydrofuran, crotonolactone, 2-5h-furanone
PubChem CID 10341
ChEBI CHEBI:38118
IUPAC Name 2H-furan-5-one
SMILES O=C1OCC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 3.0°C to 4.0°C
Color Amber or Colorless to Yellow
Density 1.1900g/mL
Boiling Point 212.0°C to 214.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.19
Solubility Information Solubility in water: insoluble
Formula Weight 84.07
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2(5H)-Furanone
Show More Show Less
compliance-icons
Product Identifier
  • 2-(5H)-Furanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.