missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Xylenesulfonic Acid, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 2,4-Xylenesulfonic Acid |
| CAS | 88-61-9 |
| Molecular Formula | C8 H10 O3 S . H2 O |
| Quantity | 2.5 g |
| Synonym | Benzenesulfonic acid','2,4-dimethyl- (9CI','ACI), 2,4-Dimethylbenzenesulfonic acid (ACI), 2,4-Xylenesulfonic acid (7CI','8CI), 2,4-Dimethylbenzene-1-sulfonic acid, m-Xylene-4-sulfonic acid, m-Xylenesulfonic acid, SXA 60 |
| InChI Formula | InChI=1S/C8H10O3S.H2O/c1-6-3-4-8(7(2)5-6)12(9,10)11;/h3-5H,1-2H3,(H,9,10,11);1H2 |
| SMILES | O.Cc1ccc(c(C)c1)S(=O)(=O)O |
| IUPAC Name | 2,4-dimethylbenzenesulfonic acid;hydrate |
| Molecular Weight (g/mol) | 204.243 |
| Formula Weight | 204.0456 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction