Learn More
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-ol, 97%, Maybridge™
Brand: Maybridge BTB09930DA
Code : 31
Additional Details : CAS Number : 103788-65-4 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
103788-65-4 | |
203.241 | |
JYWHQBLLIBQGCU-UHFFFAOYSA-N | |
725585 | |
CC1=C(N=C(O1)C2=CC=CC=C2)CCO |
C12H13NO2 | |
MFCD00100006 | |
2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethan-1-ol, 2-5-methyl-2-phenyloxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyloxazol, 4-oxazoleethanol, 5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl-ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyl-1,3-oxazole, 4-oxazoleethanol,5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl-1-ethanol | |
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethanol |
Specifications
2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)ethan-1-ol | |
1g | |
96.5 | |
97% | |
MFCD00100006 | |
JYWHQBLLIBQGCU-UHFFFAOYSA-N | |
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethanol | |
725585 | |
97% |
Amber glass bottle | |
103788-65-4 | |
100.0 | |
C12H13NO2 | |
2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethan-1-ol, 2-5-methyl-2-phenyloxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyloxazol, 4-oxazoleethanol, 5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl-ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyl-1,3-oxazole, 4-oxazoleethanol,5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl-1-ethanol | |
CC1=C(N=C(O1)C2=CC=CC=C2)CCO | |
203.241 | |
203.24 |