missing translation for 'onlineSavingsMsg'
Learn More

Fisher Chemical 2.5 LT ETHANOL SLR 96% V/V DUTY WILL BE CHARGEDat the current rate SpeciFied

Brand:  Fisher Chemical E/0551/17


2.5 LT ETHANOL SLR 96% V/V DUTY WILL BE CHARGEDat the current rate SpeciFied

Product Code. 12478720

  • ONLINE EXCLUSIVE PRICE
    £316.00
    £237.65 / 2.50 L
    Save £78.35
    24% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethanol (96% v/v)

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.