missing translation for 'onlineSavingsMsg'
Learn More

2,5-Hexanedione, 97%

Product Code. 11491707
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491707 25 g 25g
11401717 100 g 100g
11411717 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491707 Supplier Thermo Scientific Alfa Aesar Supplier No. B25686.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-13-4 | C6H10O2 | 114.144 g/mol

2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.

Solubility
Miscible with alcohol, water and diethyl ether.

Notes
Incompatible with strong oxidizing agents, strong bases, metals and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-13-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00008792
InChI Key OJVAMHKKJGICOG-UHFFFAOYSA-N
Synonym 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
PubChem CID 8035
ChEBI CHEBI:85014
IUPAC Name hexane-2,5-dione
SMILES CC(=O)CCC(=O)C

Specifications

Melting Point -6°C
Density 0.976
Boiling Point 192°C to 193°C
Flash Point 78°C (172°F)
Refractive Index 1.425
Quantity 25 g
Beilstein 506525
Merck Index 14,71
Solubility Information Miscible with alcohol,water and diethyl ether.
Formula Weight 114.15
Percent Purity 97%
Chemical Name or Material 2,5-Hexanedione
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Hexanedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2,5-Hexanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.