missing translation for 'onlineSavingsMsg'
Få mere at vide

2,5-Hexanedione, 97%

Artikelnummer. 11411717
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
25 g
100 g
500 g
Pakningsstørrelse:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11411717 500 g 500g
11491707 25 g 25g
11401717 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11411717 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B25686.36
 Restricted Item
Estimated Shipment: 30-07-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Formats
Denne vare kan ikke returneres. Se returpolicy

CAS: 110-13-4 | C6H10O2 | 114.144 g/mol

2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.

Solubility
Miscible with alcohol, water and diethyl ether.

Notes
Incompatible with strong oxidizing agents, strong bases, metals and strong reducing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 110-13-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00008792
InChI Key OJVAMHKKJGICOG-UHFFFAOYSA-N
Synonym 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
PubChem CID 8035
ChEBI CHEBI:85014
IUPAC Name hexane-2,5-dione
SMILES CC(=O)CCC(=O)C

Tekniske data

Melting Point -6°C
Density 0.976
Boiling Point 192°C to 193°C
Flash Point 78°C (172°F)
Refractive Index 1.425
Quantity 500 g
Beilstein 506525
Merck Index 14,71
Solubility Information Miscible with alcohol,water and diethyl ether.
Formula Weight 114.15
Percent Purity 97%
Chemical Name or Material 2,5-Hexanedione
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.