missing translation for 'onlineSavingsMsg'
Learn More

2-(5-formyl-2-methoxyphenoxy)acetic Acid, TRC

Product Code. 30403472
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403472 50 mg
30400126 500 mg
30404008 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403472 Supplier Toronto Research Chemicals Supplier No. TRCF65021550MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19728-22-4
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.18
SMILES COc1ccc(C=O)cc1OCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H10O5/c1-14-8-3-2-7(5-11)4-9(8)15-6-10(12)13/h2-5H,6H2,1H3,(H,12,13)
Formula Weight 210.0528
Chemical Name or Material 2-(5-formyl-2-methoxyphenoxy)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.