missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Ethynylthien-2-yl)pyridine, ≥97%, Thermo Scientific™

Product Code. 10697384
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 132464-90-5
Molecular Formula C11H7NS
Molecular Weight (g/mol) 185.244
MDL Number MFCD08271876
InChI Key CTZZEBNNWDVUFO-UHFFFAOYSA-N
Synonym 2-5-ethynylthien-2-yl pyridine, 2-5-ethynylthiophen-2-yl pyridine, pyridine,2-5-ethynyl-2-thienyl, pyridine, 2-5-ethynyl-2-thienyl, acmc-20muis, 5-ethynyl-2-2-pyridyl thiophene, 2-ethynyl-5-pyridin-2-yl thiophene
PubChem CID 15084146
IUPAC Name 2-(5-ethynylthiophen-2-yl)pyridine
SMILES C#CC1=CC=C(S1)C2=CC=CC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 185.25
Percent Purity 97%
Chemical Name or Material 2-(5-Ethynylthien-2-yl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'