missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Ethynylthien-2-yl)pyridine, ≥97%, Thermo Scientific™

Product Code. 10183484
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183484 1 g Amber Glass Bottle 1g
10697384 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10183484 Supplier Thermo Scientific Maybridge Supplier No. CC02247DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 132464-90-5
Molecular Formula C11H7NS
Molecular Weight (g/mol) 185.244
MDL Number MFCD08271876
InChI Key CTZZEBNNWDVUFO-UHFFFAOYSA-N
Synonym 2-5-ethynylthien-2-yl pyridine, 2-5-ethynylthiophen-2-yl pyridine, pyridine,2-5-ethynyl-2-thienyl, pyridine, 2-5-ethynyl-2-thienyl, acmc-20muis, 5-ethynyl-2-2-pyridyl thiophene, 2-ethynyl-5-pyridin-2-yl thiophene
PubChem CID 15084146
IUPAC Name 2-(5-ethynylthiophen-2-yl)pyridine
SMILES C#CC1=CC=C(S1)C2=CC=CC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 185.25
Percent Purity 97%
Chemical Name or Material 2-(5-Ethynylthien-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.