missing translation for 'onlineSavingsMsg'
Learn More

2,5-Ethyl Acetate Febuxostat, TRC

Product Code. 30384116
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384116 10 mg 10mg
30387967 50 mg -
30384047 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384116 Supplier Toronto Research Chemicals Supplier No. TRCE16431010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643131-93-4
Molecular Formula C20 H25 N O5 S
Molecular Weight (g/mol) 391.48
Synonym Febuxostat Impurity, Febuxostat DEE Impurity, Ethyl 2-(3-(Ethoxycarbonyl)-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylate, 2-[3-(Ethoxycarbonyl)-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester
IUPAC Name ethyl 2-(3-ethoxycarbonyl-4-isobutoxy-phenyl)-4-methyl-thiazole-5-carboxylate
SMILES CCOC(=O)c1cc(ccc1OCC(C)C)c2nc(C)c(s2)C(=O)OCC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H25NO5S/c1-6-24-19(22)15-10-14(8-9-16(15)26-11-12(3)4)18-21-13(5)17(27-18)20(23)25-7-2/h8-10,12H,6-7,11H2,1-5H3
Formula Weight 391.1453
Chemical Name or Material 2,5-Ethyl Acetate Febuxostat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.