missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diphenyloxazole, 99%, scintillation grade

Product Code. 10102770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10102770 100 g Plastic Bottle 100g
10091090 25 g Plastic Bottle 25g
10112770 500 g Plastic Bottle 500g
10793324 2.5 kg Plastic Drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10102770 Supplier Thermo Scientific Acros Supplier No. 117381000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-71-7 | C15H11NO | 221.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for fluorescence/luminescence spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-71-7
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
MDL Number MFCD00005306
InChI Key CNRNYORZJGVOSY-UHFFFAOYSA-N
Synonym PPO
PubChem CID 7105
IUPAC Name 2,5-diphenyl-1,3-oxazole
SMILES O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C to 72°C
Physical Form Crystalline Powder
CAS Min % 98.5
CAS Max % 100.0
Background < 25 cpm
Density 1.06g/mL
Color White
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Beilstein 27, 78
Packaging Plastic Bottle
Formula Weight 221.26
Percent Purity ≥98.5%
Chemical Name or Material 2, 5-Diphenyloxazole, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.