missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diphenyloxazole, Pure, Scintillation Counting, Fisher Chemical™

Product Code. 10306090 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10306090 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10306090 Supplier Fisher Chemical Supplier No. D/4865/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 92-71-7
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
MDL Number MFCD00005306
InChI Key CNRNYORZJGVOSY-UHFFFAOYSA-N
Synonym 2,5-diphenyloxazole, oxazole, 2,5-diphenyl, tritosol, 2,5-diphenyl-oxazole, ppo scintillator, dpo scintillator, usaf ek-6775, ppo van, ppo, ppo scintillator van
PubChem CID 7105
IUPAC Name 2,5-diphenyl-1,3-oxazole
SMILES O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C
Color White
Boiling Point 360°C
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 221.26g/mol
Physical Form Powder
Product Identifier
  • 2,5-Diphenyloxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.