missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(tert-butoxycarbonylamino)hexanoate (N,N'-Bis(tert-butoxycarbonyl)-L-lysine N-Succinimidyl Ester), Mikromol™
Description
2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(tert-butoxycarbonylamino)hexanoate (N,N'-Bis(tert-butoxycarbonyl)-L-lysine N-Succinimidyl Ester)
Specifications
Specifications
| Chemical Name or Material | 2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(tert-butoxycarbonylamino)hexanoate |
| CAS | 30189-36-7 |
| Analyte or Component Names | 2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(tert-butoxycarbonylamino)hexanoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H33 N3 O8 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Lysine, N2,N6-bis[(1,1-dimethylethoxy)carbonyl]-, 2,5-dioxo-1-pyrrolidinyl ester, Carbamic acid, [(1S)-1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1,5-pentanediyl]bis-, bis(1,1-dimethylethyl) ester (9CI), Carbamic acid, [1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1,5-pentanediyl]bis-, bis(1,1-dimethylethyl) ester, (S)-, Succinimide, N-[(N2,N6-dicarboxy-L-lysyl)oxy]-, di-tert-butyl ester (8CI), 42: PN: US20070066537 PAGE: 13 claimed protein, Bis(tert-butoxycarbonyl)lysine-N-hydroxysuccinimide, Nα,Nω-Di(tert-butoxycarbonyl)-L-lysine N-hydroxysuccinimide ester |
| InChI Formula | InChI=1S/C20H33N3O8/c1-19(2,3)29-17(27)21-12-8-7-9-13(22-18(28)30-20(4,5)6)16(26)31-23-14(24)10-11-15(23)25/h13H,7-12H2,1-6H3,(H,21,27)(H,22,28)/t13-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O |
| IUPAC Name | (2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?