missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylthiophenol, 98%, Thermo Scientific™

Catalog Number 11421187
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4001-61-0
C8H10S
138.228
MFCD00010020
NHAUBUMQRJWWAT-UHFFFAOYSA-N
2,5-dimethylthiophenol, 2,5-dimethyl thiophenol, benzenethiol, 2,5-dimethyl, 2,5-dimethylbenzene-1-thiol, 2,5-dimethylthiophenol;2,5-dimethylthiophenol;, p-xylenethiol, 2-mercapto-p-xylene, 2,5dimethylthiophenol, pubchem6811, 2,5-dimethyl-thiophenol
77615
2,5-dimethylbenzenethiol
CC1=CC(=C(C=C1)C)S
This item is not returnable. View return policy
4001-61-0
C8H10S
138.228
MFCD00010020
NHAUBUMQRJWWAT-UHFFFAOYSA-N
2,5-dimethylthiophenol, 2,5-dimethyl thiophenol, benzenethiol, 2,5-dimethyl, 2,5-dimethylbenzene-1-thiol, 2,5-dimethylthiophenol;2,5-dimethylthiophenol;, p-xylenethiol, 2-mercapto-p-xylene, 2,5dimethylthiophenol, pubchem6811, 2,5-dimethyl-thiophenol
77615
2,5-dimethylbenzenethiol
CC1=CC(=C(C=C1)C)S

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.02
Boiling Point 126°C to 127°C (50 mmHg)
Flash Point 83°C (181°F)
Odor Unpleasant
Refractive Index 1.5698
Quantity 5 g
UN Number UN2810
Beilstein 2040348
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylthiophenol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'