missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylthiophene, 98+%

Product Code. 11424070
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424070 25 g 25g
11444213 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424070 Supplier Thermo Scientific Alfa Aesar Supplier No. A10869.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-02-8 | C6H8S | 112.19 g/mol

2,5-Dimethylthiophene is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylthiophene is used as an intermediate in organic synthesis.

Solubility
Insoluble in water. Soluble in alcohol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-02-8
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00005452
InChI Key GWQOOADXMVQEFT-UHFFFAOYSA-N
Synonym thiophene, 2,5-dimethyl, 2,5-dimethyl-thiophene, unii-v6ddx6wb12, v6ddx6wb12, pubchem5194, 2,5 dimethylthiophene, thiophene,5-dimethyl, 2,5-dimethyl thiophene, thiophene,2,5-dimethyl, 2,5-dimethyl-2h-thiolium
PubChem CID 12514
IUPAC Name 2,5-dimethylthiophene
SMILES CC1=CC=C(S1)C

Specifications

Melting Point -63°C
Density 0.984
Boiling Point 134°C to 136°C
Flash Point 24°C (75°F)
Odor Characteristic
Refractive Index 1.512
Quantity 25 g
UN Number UN1993
Beilstein 106450
Solubility Information Insoluble in water. Soluble in alcohol,ether and benzene.
Formula Weight 112.19
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethylthiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.