missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylthiophene-3-boronic Acid, TRC

Product Code. 30374387
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374387 50 mg
30372625 100 mg
30387682 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374387 Supplier Toronto Research Chemicals Supplier No. TRCD47960850MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162607-23-0
Molecular Formula C6H9BO2S
Molecular Weight (g/mol) 156.01
Synonym B-(2,5-Dimethyl-3-thienyl)boronic Acid, (2,5-Dimethylthiophen-3-yl)boronic Acid, 2,5-Dimethyl-3-thienylboronic Acid, 2,5-Dimethylthien-3-ylboronic Acid, 2,5-Dimethylthiophene-3-boronic Acid
SMILES Cc1cc(B(O)O)c(C)s1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H9BO2S/c1-4-3-6(7(8)9)5(2)10-4/h3,8-9H,1-2H3
Chemical Name or Material 2,5-Dimethylthiophene-3-boronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.