missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethylstyrene (stabilized with 4-tert-butylcatechol), TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 2,5-dimethylstyrene |
| CAS | 2039-89-6 |
| Molecular Formula | C10 H12 |
| Quantity | 500 mg |
| Synonym | Benzene, 2-ethenyl-1,4-dimethyl-, Styrene, 2,5-dimethyl- (7CI,8CI), 2-Ethenyl-1,4-dimethylbenzene, 1,4-Dimethyl-2-ethenylbenzene, 1,4-Dimethyl-2-vinylbenzene, 2,5-Dimethylstyrene, NSC 73477 |
| InChI Formula | InChI=1S/C10H12/c1-4-10-7-8(2)5-6-9(10)3/h4-7H,1H2,2-3H3 |
| SMILES | Cc1ccc(C)c(C=C)c1 |
| IUPAC Name | 2-ethenyl-1,4-dimethylbenzene |
| Molecular Weight (g/mol) | 132.2 |
| Formula Weight | 132.0939 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction