missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylstyrene, 98%, Thermo Scientific™

Product Code. 15294854 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15294854

Brand: Thermo Scientific Acros 450820050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2039-89-6
Melting Point -35°C
Boiling Point 71°C to 72°C (13.0 hPa)
Flash Point 63°C
Assay Percent Range 97.5% min. (GC)
Molecular Formula C10H12
Refractive Index 1.5395
Quantity 5 g
Synonym 2,5-dimethylstyrene, styrene, 2,5-dimethyl, 1,4-dimethyl-2-vinylbenzene, 1,4-dimethyl-2-ethenylbenzene, benzene, 2-ethenyl-1,4-dimethyl, unii-rtm6p5k8ca, rtm6p5k8ca, styrene,5-dimethyl, acmc-1cjqw, 4-05-00-01385 beilstein handbook reference
InChI Key DBWWINQJTZYDFK-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)C)C=C
IUPAC Name 2-ethenyl-1,4-dimethylbenzene
Molecular Weight (g/mol) 132.2
PubChem CID 16265
Formula Weight 132.2
Percent Purity 98%
Chemical Name or Material 2, 5-Dimethylstyrene
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dimethylstyrene, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.