missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylstyrene, 97%, stab. with 4-tert-butylcatechol

Product Code. 11417014
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11417014

Brand: Thermo Scientific Alfa Aesar B21476.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2039-89-6 | C10H12 | 132.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-89-6
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD00008614
InChI Key DBWWINQJTZYDFK-UHFFFAOYSA-N
Synonym 2,5-dimethylstyrene, styrene, 2,5-dimethyl, 1,4-dimethyl-2-vinylbenzene, 1,4-dimethyl-2-ethenylbenzene, benzene, 2-ethenyl-1,4-dimethyl, unii-rtm6p5k8ca, rtm6p5k8ca, styrene,5-dimethyl, acmc-1cjqw, 4-05-00-01385 beilstein handbook reference
PubChem CID 16265
IUPAC Name 2-ethenyl-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)C=C

Specifications

Melting Point -35°C
Density 0.904
Boiling Point 83°C to 84°C (24 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.539
Quantity 25 g
Beilstein 2203656
Formula Weight 132.21
Percent Purity 97%
Chemical Name or Material 2,5-Dimethylstyrene, stabilized with 4-tert-butylcatechol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.