missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrrole, 97%

Product Code. 10351614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10351614 25 g 25g
10596013 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351614 Supplier Thermo Scientific Acros Supplier No. 116750250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-84-3 | C6H9N | 95.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-84-3
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.14
MDL Number MFCD00005223
InChI Key PAPNRQCYSFBWDI-UHFFFAOYSA-N
Synonym 2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole
PubChem CID 12265
IUPAC Name 2,5-dimethyl-1H-pyrrole
SMILES CC1=CC=C(N1)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange-Yellow to Yellow
Density 0.9350g/mL
Boiling Point 165.0°C (740.0 mmHg)
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5040 to 1.5070
Quantity 25 g
Beilstein 20, 172
Specific Gravity 0.935
Solubility Information Solubility in water: insoluble
Formula Weight 95.14
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2, 5-Dimethylpyrrole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethylpyrrole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.