missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenylacetic acid, 98+%, Thermo Scientific™

Product Code. 11351487
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11351487

Brand: Thermo Scientific Alfa Aesar L02022.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13612-34-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00014369
InChI Key RUSCTNYOPQOXDJ-UHFFFAOYSA-N
Synonym 2,5-dimethylphenylacetic acid, 2-2,5-dimethylphenyl acetic acid, 2,5-dimethylphenyl acetic acid, 2,5-xylylacetic acid, 2,5-dimethylphenylaceticacid, 2,5-dimethylbenzeneacetic acid, benzeneacetic acid, 2,5-dimethyl, pubchem22525, acmc-1bb4h
PubChem CID 83615
IUPAC Name 2-(2,5-dimethylphenyl)acetic acid
SMILES CC1=CC(=C(C=C1)C)CC(=O)O

Specifications

Melting Point 126°C to 130°C
Quantity 25 g
Beilstein 2502313
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethylphenylacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.