missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenylacetic acid, 98%, Thermo Scientific™

Product Code. 10472713 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10472713 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10472713 Supplier Thermo Scientific Acros Supplier No. 376620050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 128.0°C to 132.0°C
CAS 13612-34-5
CAS Min % 97.5
Color Beige
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C10H12O2
MDL Number MFCD00014369
Beilstein 09,551
Synonym 2,5-dimethylphenylacetic acid, 2-2,5-dimethylphenyl acetic acid, 2,5-dimethylphenyl acetic acid, 2,5-xylylacetic acid, 2,5-dimethylphenylaceticacid, 2,5-dimethylbenzeneacetic acid, benzeneacetic acid, 2,5-dimethyl, pubchem22525, acmc-1bb4h
InChI Key RUSCTNYOPQOXDJ-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)C)CC(=O)O
IUPAC Name 2-(2,5-dimethylphenyl)acetic acid
Molecular Weight (g/mol) 164.2
PubChem CID 83615
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 2, 5-Dimethylphenylacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.